Bismuth(III) chloride oxide, 99.95% (metals basis)
Bismuth(III) chloride oxide, 99.95% (metals basis)
Bismuth(III) chloride oxide, 99.95% (metals basis)
Thermo Scientific Chemicals

Bismuth(III) chloride oxide, 99.95% (metals basis)

CAS: 7787-59-9 | BiClHO | 261.437 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 017131.14
also known as 017131-14
Price (USD)
77.65
Special offer
Online exclusive
Ends: 28-Dec-2026
90.70
Save 13.05
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 28-Dec-2026
90.70
Save 13.05
Each
Chemical Identifiers
CAS7787-59-9
IUPAC Namechlorobismuthanone
Molecular FormulaBiClH2O
InChI KeyOMRKLWMUNWPRFB-UHFFFAOYSA-N
SMILESCl.O=[BiH]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
It is used as skin protective's, thickeners, and absorbent agents. It act as a coloring agents in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only