Tris(trimethylsilyl) phosphite, 95%
Tris(trimethylsilyl) phosphite, 95%
Tris(trimethylsilyl) phosphite, 95%
Thermo Scientific Chemicals

Tris(trimethylsilyl) phosphite, 95%

CAS: 1795-31-9 | C9H27O3PSi3 | 298.541 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 017159.04
also known as 017159-04
Price (USD)
62.65
Special offer
Online exclusive
Ends: 30-Jun-2026
73.10
Save 10.45 (14%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
62.65
Special offer
Online exclusive
Ends: 30-Jun-2026
73.10
Save 10.45 (14%)
Each
Chemical Identifiers
CAS1795-31-9
IUPAC Nametritrimethylsilyl phosphite
Molecular FormulaC9H27O3PSi3
InChI KeyVMZOBROUFBEGAR-UHFFFAOYSA-N
SMILESC[Si](C)(C)OP(O[Si](C)(C)C)O[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥94.0% (titration)
Appearance (Color)Clear colorless
FormLiquid
Tris(trimethylsilyl) phosphite is a reagent used for the mild preparation of bis(TMS) phosphonates (Arbuzow reaction). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only