Arsenic(II) sulfide, tech. 90%
Arsenic(II) sulfide, tech. 90%
Arsenic(II) sulfide, tech. 90%
Arsenic(II) sulfide, tech. 90%
Thermo Scientific Chemicals

Arsenic(II) sulfide, tech. 90%

CAS: 12044-79-0 | AsH3S | 110.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 017284.14
also known as 017284-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS12044-79-0
IUPAC Namearsanethiol
Molecular FormulaAsH3S
InChI KeyBMOFEAKQXGGUPE-UHFFFAOYSA-N
SMILESS[AsH2]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)88.1% min
Arsenic(II) sulfide is used as an primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only