Ammonium sulfite monohydrate, 92% min
Ammonium sulfite monohydrate, 92% min
Ammonium sulfite monohydrate, 92% min
Thermo Scientific Chemicals

Ammonium sulfite monohydrate, 92% min

Air Sensitive | CAS: 7783-11-1 | H10N2O4S | 134.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 017996.A7
also known as 017996-A7
Price (USD)
184.65
Online Exclusive
205.00
Save 20.35 (10%)
Each
Quantity:
5 kg
Request bulk or custom format
Price (USD)
184.65
Online Exclusive
205.00
Save 20.35 (10%)
Each
Chemical Identifiers
CAS7783-11-1
IUPAC Namediammonium hydrate sulfite
Molecular FormulaH10N2O4S
InChI KeyASPXBYAQZVXSNS-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].O.[O-]S([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or crystalline solid
Assay from Suppliers CofA≥92% (Titration)
Heavy Metals≤5ppm
Residue on ignition≤0.01%
View more
Ammonium sulfite monohydrate is used in caramel coloring in the food industry. It is also used in cosmetics as a hair straightening agent and a hair waving agent. It acts as a reducing agent in photography. Further, it is used in the production of bricks in order to use in blast furnace linings. In addition to this, it used in gas scrubbers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only