Ammonium carbamate, 98%
Ammonium carbamate, 98%
Ammonium carbamate, 98%
Thermo Scientific Chemicals

Ammonium carbamate, 98%

Ammoniating agent | CAS: 1111-78-0 | CH6N2O2 | 78.071 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 018134.30
also known as 018134-30
Price (USD)
122.65
Online Exclusive
136.00
Save 13.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
122.65
Online Exclusive
136.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS1111-78-0
IUPAC Nameammonium hydrate carbamate
Molecular FormulaCH8N2O3
InChI KeySFWLALIEAXEULE-UHFFFAOYSA-N
SMILES[NH4+].O.NC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.6% min
Ammonium carbamate is used in the preparation of fertilizers. It acts as an ammoniating agent. It plays an important role in the formation of carbamoyl phosphate, which is essential for urea cycle and pyrimidines production. In addition to this, it is used in the preparation of trimethylsilyl carbamate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only