Lithium chloride-potassium chloride
Lithium chloride-potassium chloride
Lithium chloride-potassium chloride
Thermo Scientific Chemicals

Lithium chloride-potassium chloride

CAS: 65567-96-6 | Cl2KLi | 116.938 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 018206.18
also known as 018206-18
Price (USD)
122.65
Online exclusive
136.00
Save 13.35
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
122.65
Online exclusive
136.00
Save 13.35
Each
Chemical Identifiers
CAS65567-96-6
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)61% Cl min
Lithium chloride-potassium chloride is used in separators for mixed transition-metal oxide cathodes in thermal batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only