Magnesium phosphate pentahydrate, P{2}O{5} 33%
Magnesium phosphate pentahydrate, P{2}O{5} 33%
Magnesium phosphate pentahydrate, P{2}O{5} 33%
Magnesium phosphate pentahydrate, P{2}O{5} 33%
Thermo Scientific Chemicals

Magnesium phosphate pentahydrate, P{2}O{5} 33%

CAS: 10233-87-1 | H10Mg3O13P2 | 352.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 018219.A1
also known as 018219-A1
Price (USD)
73.65
Online Exclusive
81.40
Save 7.75 (10%)
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)
73.65
Online Exclusive
81.40
Save 7.75 (10%)
Each
Add to cart
Chemical Identifiers
CAS10233-87-1
IUPAC Namemagnesium(2+) pentahydrate phosphate
Molecular FormulaH10MgO9P
InChI KeyZHDVAZVXDZNFPS-UHFFFAOYSA-K
SMILESO.O.O.O.O.[Mg++].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)P_2O_5 33% min
Magnesium phosphate pentahydrate is used in the preparation of magnesium calcium phosphate biocement(MCPB) for bone regeneration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only