Molybdenum(II) acetate dimer
Molybdenum(II) acetate dimer
Molybdenum(II) acetate dimer
Thermo Scientific Chemicals

Molybdenum(II) acetate dimer

CAS: 14221-06-8 | C8H12Mo2O8 | 428.076 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 018239.03
also known as 018239-03
Price (USD)
81.60
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
81.60
Each
Chemical Identifiers
CAS14221-06-8
IUPAC Namedimolybdenum(2+) tetraacetate
Molecular FormulaC8H12Mo2O8
InChI KeyDOOLFANBWPPEGQ-UHFFFAOYSA-J
SMILES[Mo++].[Mo++].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green
FormCrystals or powder or crystalline powder
Assay (special)Elemental Analysis: ≥98.0%
Molybdenum(II) acetate dimer is generally used as an intermediate compound in a process to form other quadruply bonded molybdenum compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only