Tin(II) 2-ethylhexanoate, tech.
Tin(II) 2-ethylhexanoate, tech.
Tin(II) 2-ethylhexanoate, tech.
Thermo Scientific Chemicals

Tin(II) 2-ethylhexanoate, tech.

CAS: 301-10-0 | C16H30O4Sn | 405.122 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 018590.A3
also known as 018590-A3
Price (USD)
279.65
Special offer
Online exclusive
Ends: 30-Jun-2026
329.00
Save 49.35 (15%)
Each
Quantity:
2 kg
Request bulk or custom format
Price (USD)
279.65
Special offer
Online exclusive
Ends: 30-Jun-2026
329.00
Save 49.35 (15%)
Each
Chemical Identifiers
CAS301-10-0
IUPAC Nameλ²-tin(2+) bis(2-ethylhexanoate)
Molecular FormulaC16H30O4Sn
InChI KeyKSBAEPSJVUENNK-UHFFFAOYNA-L
SMILES[Sn++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
Refractive Index1.4920 to 1.4960
Formliquid
Assay (Titration)Total Tin: 28.0 wt%, min
Tin(II) 2-ethylhexanoate is used as a polymerization initiator in polylactic acid production. It acts as an intermediate as well as a catalyst for urethane foams, lubricants, addition agents and stabilizers for transformer oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only