Zinc diethyldithiocarbamate, Zn 17-19.5%
Zinc diethyldithiocarbamate, Zn 17-19.5%
Zinc diethyldithiocarbamate, Zn 17-19.5%
Thermo Scientific Chemicals

Zinc diethyldithiocarbamate, Zn 17-19.5%

Chelating agent | CAS: 14324-55-1 | C10H20N2S4Zn | 361.904 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 018697.A3
also known as 018697-A3
Price (USD)
68.65
Online Exclusive
75.80
Save 7.15 (9%)
Each
Quantity:
2 kg
Request bulk or custom format
Price (USD)
68.65
Online Exclusive
75.80
Save 7.15 (9%)
Each
Specifications
CAS14324-55-1
Chemical Name or MaterialZinc diethyldithiocarbamate
ColorWhite
Concentration or Composition (by Analyte or Components)Zn 17 to 19.5%
DOT InformationTransport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
View more
Zinc diethyldithiocarbamate is a chelating ligand. It is used as a common accelerator to latex. A fast curing primary or secondary effective ultra-accelerator for natural and synthetic latex compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only