1,5-Hexadiyne, 50% in pentane
1,5-Hexadiyne, 50% in pentane
1,5-Hexadiyne, 50% in pentane
Thermo Scientific Chemicals

1,5-Hexadiyne, 50% in pentane

CAS: 628-16-0 | C6H6 | 78.114 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 018720.04
also known as 018720-04
Price (USD)
116.65
Online Exclusive
129.00
Save 12.35 (10%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
116.65
Online Exclusive
129.00
Save 12.35 (10%)
Each
Specifications
CAS628-16-0
ChEBICHEBI:37821
Chemical Name or Material1,5-Hexadiyne
Concentration or Composition (by Analyte or Components)50% in pentane
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
View more
1,5-Hexadiyne is used in the preparation of bisdehydro[12]annulene and biphenylene. It is also used as an important starting material in the preparation of (5Z,9Z)-5,9-hexadecadienoic acid. Further, it is involved in the preparation of linear organotin polymers by reaction with organotin dihydrides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only