Nickel(II) 2,4-pentanedionate, 95%
Nickel(II) 2,4-pentanedionate, 95%
Nickel(II) 2,4-pentanedionate, 95%
Thermo Scientific Chemicals

Nickel(II) 2,4-pentanedionate, 95%

Catalyst and U.V. stabilizer | CAS: 3264-82-2 | C10H14NiO4 | 256.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 018811.14
also known as 018811-14
Price (USD)
54.90
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
54.90
Each
Chemical Identifiers
CAS3264-82-2
IUPAC Namenickel(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14NiO4
InChI KeyBMGNSKKZFQMGDH-FDGPNNRMSA-L
SMILES[Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green
FormPowder or crystalline powder or crystals
Assay from Supplier's CofA≥94.0 to ≤106.0% (Titration)
Assay from Supplier's CofANickel, Ni : 21.5-24.2%
Catalyst and U.V. stabilizer.Nickel(II) 2,4-pentanedionate is used as a precursor to nickel bis(cyclooctadiene) catalyst. It is also used in the deposition of nickel(II) oxide thin film by sol-gel techniques on conductive glass substrates. Further, it is used in organic synthesis to produce organometals. It is associated with dimethylgold(III) acetylacetonate is used in gold on nickel plating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only