Iron(III) meso-tetraphenylporphine chloride
Iron(III) meso-tetraphenylporphine chloride
Iron(III) meso-tetraphenylporphine chloride
Thermo Scientific Chemicals

Iron(III) meso-tetraphenylporphine chloride

CAS: 16456-81-8 | C44H30Cl3FeN4 | 776.947 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 018829.MD
also known as 018829-MD
Price (USD)
52.65
Special offer
Online exclusive
Ends: 30-Jun-2026
61.80
Save 9.15 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
52.65
Special offer
Online exclusive
Ends: 30-Jun-2026
61.80
Save 9.15 (15%)
Each
Specifications
CAS16456-81-8
Chemical Name or MaterialIron(III) meso-tetraphenylporphine chloride
Formula Weight704.04
IUPAC Nameiron(3+);5,10,15,20-tetraphenyl-21,22-dihydroporphyrin;trichloride
InChI KeyQDTNPIPZGCOFPA-UHFFFAOYSA-K
View more
Iron(III) meso-tetraphenylporphine chloride is used catalyzing reagent for silylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only