Diiodobis(triphenylphosphine)nickel(II)
Diiodobis(triphenylphosphine)nickel(II)
Diiodobis(triphenylphosphine)nickel(II)
Thermo Scientific Chemicals

Diiodobis(triphenylphosphine)nickel(II)

CAS: 82308-17-6 | C36H30I2NiP2 | 837.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 018869.04
also known as 018869-04
Price (USD)
58.40
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
58.40
Each
Specifications
CAS82308-17-6
Chemical Name or MaterialDiiodobis(triphenylphosphine)nickel(II)
ColorBrown
Formula Weight837.08
Health Hazard 1H317-H350
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only