Copper(I) bromide-dimethyl sulfide complex
Copper(I) bromide-dimethyl sulfide complex
Copper(I) bromide-dimethyl sulfide complex
Thermo Scientific Chemicals

Copper(I) bromide-dimethyl sulfide complex

CAS: 54678-23-8 | C2H6BrCuS | 205.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 019368.04
also known as 019368-04
Price (USD)
28.60
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
28.60
Each
Chemical Identifiers
CAS54678-23-8
IUPAC Name(methylsulfanyl)methane; bromocopper
Molecular FormulaC2H6BrCuS
InChI KeyPMHQVHHXPFUNSP-UHFFFAOYSA-M
SMILES[Cu]Br.CSC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Iodometric Titration)> 97.0%
Appearance (Color)White to off-white to faint green
Formpowder
Copper(I) bromide-dimethyl sulfide complex is used as catalyst for adding Grignard reagents to fullerenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only