2-Methylbutane, 99+%
2-Methylbutane, 99+%
2-Methylbutane, 99+%
Thermo Scientific Chemicals

2-Methylbutane, 99+%

CAS: 78-78-4 | C5H12 | 72.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
4 x 500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 019387.AP
also known as 019387-AP
Price (USD)
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS78-78-4
IUPAC Name2-methylbutane
Molecular FormulaC5H12
InChI KeyQWTDNUCVQCZILF-UHFFFAOYSA-N
SMILESCCC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥99.0%
FormLiquid
2-Methylbutane is used as a chemical intermediate. It acts as a blowing agent for polystyrene and gasoline additive. It is used as a solvent of polyethylene and involved in the preparation of polystyrene foam and polyurethane foam. Further, it is used in a closed loop in geothermal power production to operate turbines. In addition to this, it is used to freeze biological samples like tissues in place of dry ice for cryosectioning in histology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only