Xylenes, Semiconductor Grade
Xylenes, Semiconductor Grade
Xylenes, Semiconductor Grade
Thermo Scientific Chemicals

Xylenes, Semiconductor Grade

CAS: 1330-20-7 | (CH3)2C6H4
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
Catalog number 019402.M1
also known as 019402-M1
Price (USD)
212.00
Each
Quantity:
4 x 1 L
Request bulk or custom format
Price (USD)
212.00
Each
Chemical Identifiers
CAS1330-20-7
IUPAC Name1,3-xylene
Molecular FormulaC8H10
InChI KeyIVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Trace MetalZinc, Zn: ≤0.1ppm (≤100ppb)
Impurity contentBoron, B: ≤0.1ppm (≤100ppb)
Water Content (Karl Fischer Titration)≤0.02%
Trace MetalAluminum, Al: ≤0.1ppm (≤100ppb)
Trace MetalArsenic, As, and Antimony, Sb (as Arsenic); ≤0.01ppm (≤10ppb)
View more
Xylene is used as a solvent in pharmaceutical, printing, rubber, and leather industries. It is an important component of ink, rubber and adhesives as well as in thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels, and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate. It acts as a cleaning agent for steel, silicon wafers and integrated circuits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only