Zinc iron oxide
Zinc iron oxide
Zinc iron oxide
Thermo Scientific Chemicals

Zinc iron oxide

CAS: 12063-19-3 | Fe2H2O4Zn | 243.082 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 019407.22
also known as 019407-22
Price (USD)
351.65
Special offer
Online exclusive
Ends: 30-Jun-2026
414.00
Save 62.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
351.65
Special offer
Online exclusive
Ends: 30-Jun-2026
414.00
Save 62.35 (15%)
Each
Chemical Identifiers
CAS12063-19-3
IUPAC Namebis(ironoylol) zinc
Molecular FormulaFe2H2O4Zn
InChI KeyNNGHIEIYUJKFQS-UHFFFAOYSA-L
SMILES[Zn].O[Fe]=O.O[Fe]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity content1% max
Zinc iron oxide is used as pigments, especially in applications related to heat stability. It is also used in high corrosion-resistant coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only