Copper(II) acetate, anhydrous
Copper(II) acetate, anhydrous
Copper(II) acetate, anhydrous
Thermo Scientific Chemicals

Copper(II) acetate, anhydrous

CAS: 142-71-2 | C4H6CuO4 | 181.634 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 019417.22
also known as 019417-22
Price (USD)
163.65
Special offer
Online exclusive
Ends: 30-Jun-2026
192.00
Save 28.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
163.65
Special offer
Online exclusive
Ends: 30-Jun-2026
192.00
Save 28.35 (15%)
Each
Chemical Identifiers
CAS142-71-2
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97% min
Copper(II) acetate is used in biochemical applications such as DNA extraction. It is used as a source of copper in inorganic synthesis, an oxidizing agent and catalyst in organic synthesis. It is also used to couple two terminal alkynes to make a 1,3-diyne. It is widely used in pigments, manufacture of paris green, textile dyeing, skin conditioner in cosmetics, antioxidant and stabilizer in food grade polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only