1-Cyclohexene-1-acetic acid
1-Cyclohexene-1-acetic acid
1-Cyclohexene-1-acetic acid
Thermo Scientific Chemicals

1-Cyclohexene-1-acetic acid

CAS: 18294-87-6 | C8H12O2 | 140.182 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 019462.14
also known as 019462-14
Price (USD)
333.65
Special offer
Online exclusive
Ends: 30-Jun-2026
392.00
Save 58.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
333.65
Special offer
Online exclusive
Ends: 30-Jun-2026
392.00
Save 58.35 (15%)
Each
Specifications
CAS18294-87-6
Chemical Name or Material1-Cyclohexene-1-acetic acid
Density1.04 g/mL
EINECS Number242-172-1
Formula Weight140.18
View more
1-Cyclohexene-1-acetic acid used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only