Iron(III) hydroxide, alpha-phase
Iron(III) hydroxide, alpha-phase
Iron(III) hydroxide, alpha-phase
Thermo Scientific Chemicals

Iron(III) hydroxide, alpha-phase

For Use in Purifying Water, As Absorbent in Chemical Processes, As a Pigment | CAS: 20344-49-4 | FeHO2 | 88.851 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 019496.14
also known as 019496-14
Price (USD)
655.65
Online Exclusive
728.00
Save 72.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
655.65
Online Exclusive
728.00
Save 72.35 (10%)
Each
Chemical Identifiers
CAS20344-49-4
IUPAC Nameironoylol
Molecular FormulaFeHO2
InChI KeyAEIXRCIKZIZYPM-UHFFFAOYSA-M
SMILESO[Fe]=O
View more
SpecificationsSpecification SheetSpecification Sheet
X-Ray Diffraction (XRD)Matches reference
Iron(III) hydroxide is used in purifying water and as an absorbent in chemical processes. It is also used as a pigment and as a catalyst. Further, it is used in some tattoo ink and in aquarium water treatment as a phosphate binder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only