Samco™ Narrow Mouth Bio-Tite™ 90mL (3 oz.) 48mm Specimen Containers
Samco™ Narrow Mouth Bio-Tite™ 90mL (3 oz.) 48mm Specimen Containers
Samco™ Narrow Mouth Bio-Tite™ 90mL (3 oz.) 48mm Specimen Containers
Samco™ Narrow Mouth Bio-Tite™ 90mL (3 oz.) 48mm Specimen Containers
Thermo Scientific™

Samco™ Narrow Mouth Bio-Tite™ 90mL (3 oz.) 48mm Specimen Containers

Prevent bio-hazardous spills while conserving laboratory space with these stackable containers, featuring a proprietary, all plastic, leak-resistant seal.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityLabelClosure ColorNo. per Case
020038
also known as 02 0038
SterileTabbedNatural400/Cs.
020016
also known as 02 0016
SterileTabbedOrange400/Cs. (bulk)
020214
also known as 02 0214
SterileTabbedOrange400/Cs.
020039
also known as 02 0039
SterileTabbedWhite400/Cs.
020146
also known as 02 0146
Non-sterileNo LabelYellow400/Cs.
020148
also known as 02 0148
Non-sterileNon-tabbedYellow400/Cs.
Catalog number 020038
also known as 02 0038
Price (USD)
-
Sterility:
Sterile
Label:
Tabbed
Closure Color:
Natural
No. per Case:
400/Cs.
Prevent bio-hazardous spills while conserving laboratory space with stackable Thermo Scientific™ Samco™ Narrow-Mouth Bio-Tite™ 90mL (3 oz.) 48mm Specimen Containers. These shatter-resistant containers feature a proprietary, all plastic, leak-resistant seal. They are tested at varying pressures, making them ideal for use in various transport systems.
Features:
  • Dual-threaded caps create leak-resistant seal for superior safety and handling
  • Top and bottom positive grip knurls are easy to grip and open using latex gloves
  • Virgin polypropylene allows higher clarity to see samples, and will not become brittle
  • Sterile indicator tab assures tamper evidence
  • Bilingual English and Spanish label facilitates accurate usage

Ordering Information:

  • Available nonsterile or gamma irradiation sterilized
  • Available labeled or non-labeled
  • Available bulk or individually packed
  • In cases of 400
Specifications
Capacity (Metric)90 mL
Closure ColorNatural
Closure Size48 mm
Closure TypeBio-Tite Lid
DescriptionSterile; Tabbed label; Natural caps
Diameter (Metric)48 mm
LabelTabbed
No. per Case400/Cs.
Closure IncludedYes
MaterialVirgin Polypropylene
Product TypeSpecimen Container
ShapeCylindrical
SterilitySterile
Unit SizeCase of 400