Potassium trifluoromethanesulfonate, 98%
Potassium trifluoromethanesulfonate, 98%
Potassium trifluoromethanesulfonate, 98%
Thermo Scientific Chemicals

Potassium trifluoromethanesulfonate, 98%

CAS: 2926-27-4 | CF3KO3S | 188.162 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 020361.18
also known as 020361-18
Price (USD)
159.65
Special offer
Online exclusive
Ends: 30-Jun-2026
188.00
Save 28.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
159.65
Special offer
Online exclusive
Ends: 30-Jun-2026
188.00
Save 28.35 (15%)
Each
Chemical Identifiers
CAS2926-27-4
IUPAC Namepotassium trifluoromethanesulfonate
Molecular FormulaCF3KO3S
InChI KeyGLGXXYFYZWQGEL-UHFFFAOYSA-M
SMILES[K+].[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Purity≥97.5%
FormPowder or crystals or crystalline powder
Potassium trifluoromethanesulfonate is used as a reagent in the synthesis of guanine-quadruplex hybrid materials. It acts as a supporting electrolyte in the electrochemical study of evidence for gold anion in ethylenediamine. It is involved in the preparation of N-fluoro-2,4,6-trimethylpyridinium triflate by reacting with 2,4,6-trimethyl-pyridine. Further, it is used in the preparation of imidazolium salt, 3-methyl-1-(3R,3aR,6S,6aR)-[6-(benzyloxy)-hexahydrofuro[3,2-b]furan-3-yl]imidazolium trifluoromethanesulfonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only