Methyl malonyl chloride, 97%
Methyl malonyl chloride, 97%
Methyl malonyl chloride, 97%
Thermo Scientific Chemicals

Methyl malonyl chloride, 97%

CAS: 37517-81-0 | C4H5ClO3 | 136.531 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 020823.06
also known as 020823-06
Price (USD)
62.80
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
62.80
Each
Chemical Identifiers
CAS37517-81-0
IUPAC Namemethyl 3-chloro-3-oxopropanoate
Molecular FormulaC4H5ClO3
InChI KeyUTBCRHAMJFMIIR-UHFFFAOYSA-N
SMILESCOC(=O)CC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
FormLiquid
Refractive Index1.4310-1.4350 @ 20°C
Methyl malonyl chloride is used as pharmaceutical intermediate. Used in anti-cancer drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only