Sodium trifluoromethanesulfonate
Sodium trifluoromethanesulfonate
Sodium trifluoromethanesulfonate
Thermo Scientific Chemicals

Sodium trifluoromethanesulfonate

CAS: 2926-30-9 | CF3NaO3S | 172.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 020837.04
also known as 020837-04
Price (USD)
30.20
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
30.20
Each
Chemical Identifiers
CAS2926-30-9
IUPAC Namesodium trifluoromethanesulfonate
Molecular FormulaCF3NaO3S
InChI KeyXGPOMXSYOKFBHS-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Purity≥95.0%
Assay (unspecified)Sodium (Na): ≥12.7%
FormPowder
Catalyzes the aza Diels-Alder reaction of Danishefsky's diene with imines in water under neutral conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only