Cerium(III) chloride, anhydrous, 99.5% (REO)
Cerium(III) chloride, anhydrous, 99.5% (REO)
Cerium(III) chloride, anhydrous, 99.5% (REO)
Thermo Scientific Chemicals

Cerium(III) chloride, anhydrous, 99.5% (REO)

For Incandescent Gas Mantles, Spectrography, Polymerization of Olefins | CAS: 7790-86-5 | CeCl3 | 246.466 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 021112.09
also known as 021112-09
Price (USD)
77.65
Special offer
Online exclusive
Ends: 28-Dec-2026
91.50
Save 13.85
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 28-Dec-2026
91.50
Save 13.85
Each
Chemical Identifiers
CAS7790-86-5
IUPAC Namecerium(3+) trichloride
Molecular FormulaCeCl3
InChI KeyVYLVYHXQOHJDJL-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Ce+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Total Rare Earth Oxide Impurities≤0.5% (5000 ppm)
Cerium(III) chloride is used in incandescent gas mantles, spectrography and polymerization of olefins. It acts as a Lewis acid and used in Friedel-Crafts alkylation and acylation reactions in organic synthesis. It is used as a precursor for the preparation of other cerium salts, such as cerium(III) trifluoromethanesulfonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only