Cerium(III) 2,4-pentanedionate hydrate
Cerium(III) 2,4-pentanedionate hydrate
Cerium(III) 2,4-pentanedionate hydrate
Thermo Scientific Chemicals

Cerium(III) 2,4-pentanedionate hydrate

CAS: 206996-61-4 | C15H21CeO6 | 437.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 021116.22
also known as 021116-22
Price (USD)
191.65
Online exclusive
213.00
Save 21.35
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
191.65
Online exclusive
213.00
Save 21.35
Each
Chemical Identifiers
CAS206996-61-4
IUPAC Namecerium(3+) tris(2,4-dioxopentan-3-ide)
Molecular FormulaC15H21CeO6
InChI KeyLCRQZRNVLFDPAZ-UHFFFAOYSA-N
SMILES[Ce+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to brown
FormPowder or crystals or crystalline powder
Assay27-32% Ce
Assay (unspecified)27-32% Ce
Cerium(III) 2,4-pentanedionate hydrate used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only