Calcium peroxide, typically 65%
Calcium peroxide, typically 65%
Calcium peroxide, typically 65%
Calcium peroxide, typically 65%
Thermo Scientific Chemicals

Calcium peroxide, typically 65%

Rubber stabilizer | CAS: 1305-79-9 | CaO2 | 72.076 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 021157.36
also known as 021157-36
Price (USD)
168.65
Special offer
Online exclusive
Ends: 30-Jun-2026
198.00
Save 29.35 (15%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
168.65
Special offer
Online exclusive
Ends: 30-Jun-2026
198.00
Save 29.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS1305-79-9
IUPAC Namecalcium dioxidanediide
Molecular FormulaCaO2
InChI KeyLHJQIRIGXXHNLA-UHFFFAOYSA-N
SMILES[Ca++].[O-][O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)65% min
Calcium peroxide is used as a rubber stabilizer, used in different areas of industry and agriculture. In agriculture it is used as an oxygen fertilizer, and is also used in the presowing treatments of rice seed. It is used as flour bleaching agent and improving agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only