Tin(II) acetate, 95%
Tin(II) acetate, 95%
Tin(II) acetate, 95%
Tin(II) acetate, 95%
Thermo Scientific Chemicals

Tin(II) acetate, 95%

CAS: 638-39-1 | C4H6O4Sn | 236.798 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 022361.14
also known as 022361-14
Price (USD)
271.65
Online Exclusive
302.00
Save 30.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
271.65
Online Exclusive
302.00
Save 30.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS638-39-1
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)94.1% min
Tin(II) acetate is used as a precursor in the production of tin anode, Sn-Cu bimetallic nanoparticles, high surface area tin oxide catalysts. It acts as Lewis acid catalysts for the esterification and transesterification reactions. It serves as an efficient catalytic system for carbon-hydrogen activation of methoxy group of anisole in presence of palladium(II) acetate. It plays an important role in stannous oral care compositions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only