Product Image
Thermo Scientific™
DRI™ Barbiturate Drugs of Abuse Assays
Achieve high analytical accuracy and eliminate time-consuming steps in reagent preparation with ready-to-use assays in liquid form.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
02261 x 500 mL
02251 x 100 mL
100156483 x 18 mL
Catalog number 0226
Price (USD)
-
Quantity:
1 x 500 mL
Achieve high analytical accuracy and eliminate time-consuming steps in reagent preparation with ready-to-use assays in liquid form. Thermo Scientific™ DRI™ Barbiturate Drugs of Abuse Assays have been field-tested on a wide range of analyzers, and they are enthusiastically accepted in laboratories where cost-control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Multi-Drug Controls
DescriptionDRI Barbiturate Drugs of Abuse Assays
Quantity1 x 500 mL
Sample TypeUrine
TypeReagent
Detectable AnalytesSecobarbital
DoA CalibratorsMulti-drug Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C