Cadmium arsenide, 99% (metals basis), Thermo Scientific Chemicals
Cadmium arsenide, 99% (metals basis), Thermo Scientific Chemicals
Cadmium arsenide, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium arsenide, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
022773.09
also known as 022773-09
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 022773.09
also known as 022773-09
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS12006-15-4
IUPAC Namebis(cadmio-λ²-arsanyl) cadmium
Molecular FormulaAs2Cd3
InChI KeyFSIONULHYUVFFA-UHFFFAOYSA-N
SMILES[Cd].[As]=[Cd].[As]=[Cd]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Cadmium arsenide has a electronic properties and is used in 3D crystal structure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only