Ethanolamine, 99%, H<sub>2</sub>O 0.5% max, Thermo Scientific Chemicals
Ethanolamine, 99%, H<sub>2</sub>O 0.5% max, Thermo Scientific Chemicals
Ethanolamine, 99%, H<sub>2</sub>O 0.5% max, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethanolamine, 99%, H2O 0.5% max, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
022793.30
also known as 022793-30
250 g
Catalog number 022793.30
also known as 022793-30
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
50.50
Save 7.85 (16%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
50.50
Save 7.85 (16%)
Each
Chemical Identifiers
CAS141-43-5
IUPAC Name2-aminoethan-1-ol
Molecular FormulaC2H7NO
InChI KeyHZAXFHJVJLSVMW-UHFFFAOYSA-N
SMILESNCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear
Color (APHA)15 max
Assay from Suppliers CofA≥98.5%
Water Content (Karl Fischer Titration)≤0.5%
FormLiquid
Ethanolamine is used as a surfactant, fluorimetric reagent, and to remove CO2 and H2S from natural gas and other gases, functional fluids, intermediates. It is also used as feedstock in the production of detergents, emulsifiers, polishes, pharmaceuticals, corrosion inhibitors, chemical intermediates, ion exchange agents, plating agents and surface treating agents, electrical and electronic products, paper products, personal care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only