Cyclohexane, ACS, 99+%
Cyclohexane, ACS, 99+%
Cyclohexane, ACS, 99+%
Thermo Scientific Chemicals

Cyclohexane, ACS, 99+%

Maximum level of impurities: Color (APHA) 10 Evaporation residue 0.002%, Substances darkened by sulfuric acid P.T., H2O 0.02% | CAS: 110-82-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
4 x 1 L
Catalog number 022864.M1
also known as 022864-M1
Price (USD)
175.65
Special offer
Online exclusive
Ends: 30-Jun-2026
207.00
Save 31.35 (15%)
Each
Quantity:
4 x 1 L
Request bulk or custom format
Price (USD)
175.65
Special offer
Online exclusive
Ends: 30-Jun-2026
207.00
Save 31.35 (15%)
Each
Chemical Identifiers
CAS110-82-7
IUPAC Namecyclohexane
Molecular FormulaC6H12
InChI KeyXDTMQSROBMDMFD-UHFFFAOYSA-N
SMILESC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Color (APHA)≤10
Assay (GC)≥99.0% (corrected for water)
Residue on Evaporation≤0.002%
View more
Cyclohexane is used in environmental and GC analysis; for industrial production of adipic acid and caprolactam, which is a precursor of nylon and as a solvent, and to make nylon and paint remover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
TD&R FAM