Citric acid monohydrate, 99.5+%
Citric acid monohydrate, 99.5+%
Citric acid monohydrate, 99.5+%
Thermo Scientific Chemicals

Citric acid monohydrate, 99.5+%

CAS: 5949-29-1 | C6H10O8 | 210.138 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Catalog number 022869.22
also known as 022869-22
Price (USD)
33.65
Special offer
Online exclusive
Ends: 30-Jun-2026
39.80
Save 6.15 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 30-Jun-2026
39.80
Save 6.15 (15%)
Each
Chemical Identifiers
CAS5949-29-1
IUPAC Name2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
Molecular FormulaC6H10O8
InChI KeyYASYEJJMZJALEJ-UHFFFAOYSA-N
SMILESO.OC(=O)CC(O)(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on ignition0.05% max
Assay (Aqueous acid-base Titration)>/= 99.5%
Moisture holding capacity
Trace Metals
Citric acid monohydrate is used in the preparation of citrate buffer in platelets for intravital microscopy. It acts as a pH-control agent in foods, beverages and pharmaceuticals applications. It acts as an iron chelator. In animals, it improves the utilization of nutritional calcium. It is a useful buffer component for antigen and epitope unmasking. It is also used as an acidifier, flavoring agent and chelating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only