Nickel(II) carbonate, anhydrous, 98%
Nickel(II) carbonate, anhydrous, 98%
Nickel(II) carbonate, anhydrous, 98%
Nickel(II) carbonate, anhydrous, 98%
Thermo Scientific Chemicals

Nickel(II) carbonate, anhydrous, 98%

CAS: 3333-67-3 | CNiO3 | 118.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 022897.18
also known as 022897-18
Price (USD)
53.65
Special offer
Online exclusive
Ends: 30-Jun-2026
62.80
Save 9.15 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 30-Jun-2026
62.80
Save 9.15 (15%)
Each
Chemical Identifiers
CAS3333-67-3
IUPAC Namenickel(2+) carbonate
Molecular FormulaCNiO3
InChI KeyZULUUIKRFGGGTL-UHFFFAOYSA-L
SMILES[Ni++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale green
FormCrystals or crystalline powder or powder
Assay from Suppliers CofA≥97.5%
Nickel(II) carbonate is used in sulphamate baths, metal phosphating, electroplating and ceramic applications. It acts as a precursor to catalysts and an intermediate in the hydrometallurgical purification of nickel from its ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only