Potassium molybdenum oxide, anhydrous, 99.8% (metals basis)
Potassium molybdenum oxide, anhydrous, 99.8% (metals basis)
Potassium molybdenum oxide, anhydrous, 99.8% (metals basis)
Thermo Scientific™

Potassium molybdenum oxide, anhydrous, 99.8% (metals basis)

CAS: 13446-49-6 | K2MoO4 | 238.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 022898.09
also known as 022898-09
Price (USD)
44.65
Special offer
Online exclusive
Ends: 28-Dec-2026
52.40
Save 7.75
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 28-Dec-2026
52.40
Save 7.75
Each
Chemical Identifiers
CAS13446-49-6
IUPAC Namedipotassium dioxomolybdenumbis(olate)
Molecular FormulaK2MoO4
InChI KeyNUYGZETUIUXJCO-UHFFFAOYSA-N
SMILES[K+].[K+].[O-][Mo]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
Potassium molybdenum oxide used in chemical research, Chemicals, pharmaceutical research, pharmaceutical intermediates reagents, Laboratory chemicals, Manufacture of substances. And it is an important raw material and intermediates used in organic synthesis, agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only