2,2,4-Trimethylpentane, HPLC Grade, 99.7+%
2,2,4-Trimethylpentane, HPLC Grade, 99.7+%
2,2,4-Trimethylpentane, HPLC Grade, 99.7+%
Thermo Scientific Chemicals

2,2,4-Trimethylpentane, HPLC Grade, 99.7+%

Maximum level of impurities: Evaporation residue 3ppm, Chlorinated pesticides 5ppt, H2O 0.02% | CAS: 540-84-1 | C8H18 | 114.232 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
Catalog number 022901.K7
also known as 022901-K7
Price (USD)
247.00
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
247.00
Each
Chemical Identifiers
CAS540-84-1
IUPAC Name2,2,4-trimethylpentane
Molecular FormulaC8H18
InChI KeyNHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILESCC(C)CC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at205nm: ≤1.00 AU
Optical Absorbance at230nm: ≤0.10 au
Residue on Evaporation≤3 ppm
Appearance (Color)Colorless
Assay (GC)≥99.7%
View more
2,2,4-Trimethylpentane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
RUO – Research Use Only