o-Xylene, HPLC Grade, 96% min
o-Xylene, HPLC Grade, 96% min
o-Xylene, HPLC Grade, 96% min
Thermo Scientific Chemicals

o-Xylene, HPLC Grade, 96% min

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.04% | CAS: 95-47-6 | C8H10 | 106.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
Catalog number 022902.K7
also known as 022902-K7
Price (USD)
277.65
Special offer
Online exclusive
Ends: 30-Jun-2026
326.00
Save 48.35 (15%)
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
277.65
Special offer
Online exclusive
Ends: 30-Jun-2026
326.00
Save 48.35 (15%)
Each
Chemical Identifiers
CAS95-47-6
IUPAC Name1,2-xylene
Molecular FormulaC8H10
InChI KeyCTQNGGLPUBDAKN-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
CommentSub micron filtered
Optical Absorbance at300nm: ≤0.2 AU
Appearance (Color)Colorless
Water Content (Karl Fischer Titration)≤0.04%
View more
o-Xylene is largely used in the production of phthalic anhydride, and is generally extracted by distillation from a mixed Xylene stream in a plant primarily designed for p-Xylene production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only