Isobutanol, HPLC Grade, 99+%
Isobutanol, HPLC Grade, 99+%
Isobutanol, HPLC Grade, 99+%
Thermo Scientific Chemicals

Isobutanol, HPLC Grade, 99+%

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.05% | CAS: 78-83-1 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
Catalog number 022908.K7
also known as 022908-K7
Price (USD)
166.65
Special offer
Online exclusive
Ends: 30-Jun-2026
196.00
Save 29.35 (15%)
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
166.65
Special offer
Online exclusive
Ends: 30-Jun-2026
196.00
Save 29.35 (15%)
Each
Chemical Identifiers
CAS78-83-1
IUPAC Name2-methylpropan-1-ol
Molecular FormulaC4H10O
InChI KeyZXEKIIBDNHEJCQ-UHFFFAOYSA-N
SMILESCC(C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on Evaporation5ppm
Residual water0.05%
Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is used as a mobile phase in High Performance Liquid Chromatography. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful for the production of renewable chemicals and fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only