n-Heptane, HPLC grade, 99+%
n-Heptane, HPLC grade, 99+%
n-Heptane, HPLC grade, 99+%
Thermo Scientific™

n-Heptane, HPLC grade, 99+%

CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
4 L
4 x 4 L
Catalog number 022911.K2
also known as 022911-K2
Price (USD)
85.10
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
85.10
Each
Chemical Identifiers
CAS142-82-5
IUPAC Nameheptane
Molecular FormulaC7H16
InChI KeyIMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILESCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤0.02% [US & UK]
Appearance (Color)Clear colorless
Assay (GC)≥99.0%
Residue on Evaporation≤0.00030% (3.0ppm) [US & UK]
Optical Absorbance at195nm: ≥10% transmittance [UK]
View more
n-Heptane is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
RUO – Research Use Only