N,N-Dimethylformamide, HPLC Grade, 99.7+%
N,N-Dimethylformamide, HPLC Grade, 99.7+%
N,N-Dimethylformamide, HPLC Grade, 99.7+%
Thermo Scientific Chemicals

N,N-Dimethylformamide, HPLC Grade, 99.7+%

CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
6 x 1 L
Catalog number 022915.M6
also known as 022915-M6
Price (USD)
656.65
Online Exclusive
729.00
Save 72.35 (10%)
Each
Quantity:
4 x 4 L
Request bulk or custom format
Price (USD)
656.65
Online Exclusive
729.00
Save 72.35 (10%)
Each
Chemical Identifiers
CAS68-12-2
IUPAC NameN,N-dimethylformamide
Molecular FormulaC3H7NO
InChI KeyZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILESCN(C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water0.05%
Residue on Evaporation5ppm
N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films. It is used as a mobile phase in High Performance Liquid Chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only