1,2-Dichloroethane, HPLC Grade, 99% min
1,2-Dichloroethane, HPLC Grade, 99% min
1,2-Dichloroethane, HPLC Grade, 99% min
Thermo Scientific Chemicals

1,2-Dichloroethane, HPLC Grade, 99% min

CAS: 107-06-2 | C2H4Cl2 | 98.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 022918.K2
also known as 022918-K2
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS107-06-2
IUPAC Name1,2-dichloroethane
Molecular FormulaC2H4Cl2
InChI KeyWSLDOOZREJYCGB-UHFFFAOYSA-N
SMILESClCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.0%
Residual water≤0.02%
FormLiquid
Appearance (Color)Clear colorless
Color (APHA)≤10
View more
1,2-Dichloroethane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only