Chlorobenzene, Spectrophotometric Grade, 99.9%
Chlorobenzene, Spectrophotometric Grade, 99.9%
Chlorobenzene, Spectrophotometric Grade, 99.9%
Thermo Scientific Chemicals

Chlorobenzene, Spectrophotometric Grade, 99.9%

CAS: 108-90-7 | C6H5Cl | 112.556 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
4 x 1 L
Catalog number 022921.K7
also known as 022921-K7
Price (USD)
296.65
Special offer
Online exclusive
Ends: 30-Jun-2026
349.00
Save 52.35 (15%)
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
296.65
Special offer
Online exclusive
Ends: 30-Jun-2026
349.00
Save 52.35 (15%)
Each
Chemical Identifiers
CAS108-90-7
IUPAC Namechlorobenzene
Molecular FormulaC6H5Cl
InChI KeyMVPPADPHJFYWMZ-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at350nm: ≤0.02AU [US]
Optical Absorbance at290nm: ≥50% transmittance [UK]
Optical Absorbance at295nm: ≥80% transmittance [UK]
Optical Absorbance at370-400nm: ≥99% transmittance [UK]
Optical Absorbance at400nm: ≤0.005AU
View more
Chlorobenzene is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only