1-Propanol, Spectrophotometric Grade, 99%
1-Propanol, Spectrophotometric Grade, 99%
1-Propanol, Spectrophotometric Grade, 99%
Thermo Scientific Chemicals

1-Propanol, Spectrophotometric Grade, 99%

CAS: 71-23-8 | C3H8O | 60.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Catalog number 022933.K2
also known as 022933-K2
Price (USD)
79.90
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
79.90
Each
Chemical Identifiers
CAS71-23-8
IUPAC Namepropan-1-ol
Molecular FormulaC3H8O
InChI KeyBDERNNFJNOPAEC-UHFFFAOYSA-N
SMILESCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on Evaporation4ppm
Residual water0.08%
1-Propanol is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only