Vanadium(III) 2,4-pentanedionate, typically 97%
Vanadium(III) 2,4-pentanedionate, typically 97%
Vanadium(III) 2,4-pentanedionate, typically 97%
Thermo Scientific Chemicals

Vanadium(III) 2,4-pentanedionate, typically 97%

CAS: 13476-99-8 | C15H21O6V | 348.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 022940.14
also known as 022940-14
Price (USD)
85.30
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
85.30
Each
Chemical Identifiers
CAS13476-99-8
IUPAC Namevanadium(3+) tris(2,4-dioxopentan-3-ide)
Molecular FormulaC15H21O6V
InChI KeyZVFSRLXFKIWGRT-UHFFFAOYSA-N
SMILES[V+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green to brown
Assay (unspecified)Vanadium (V): ≥14.0% (≥ 96.0% purity calc.)
FormPowder
Vanadium(III) 2,4-pentanedionate is used as homogeneous catalyst, primary and secondary intermediates, Laboratory chemicals, and manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only