Trimethyl borate, 99.9995+% (metals basis)
Trimethyl borate, 99.9995+% (metals basis)
Trimethyl borate, 99.9995+% (metals basis)
Trimethyl borate, 99.9995+% (metals basis)
Thermo Scientific Chemicals

Trimethyl borate, 99.9995+% (metals basis)

CAS: 121-43-7 | C3H9BO3 | 103.912 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 022975.30
also known as 022975-30
Price (USD)
261.65
Online Exclusive
290.00
Save 28.35 (10%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
261.65
Online Exclusive
290.00
Save 28.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS121-43-7
IUPAC Nametrimethyl borate
Molecular FormulaC3H9BO3
InChI KeyWRECIMRULFAWHA-UHFFFAOYSA-N
SMILESCOB(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.9995% (metals basis)
Impurity contentNa: ≤5ppm
Trimethyl borate is a useful reagent in organic synthesis. It is involved in the production of resins, waxes and paints and acts as a methylation agent. As a boron source, it is used to prepare flame retardants, anti-oxidants and corrosion inhibitors. It reacts with Grignard reagents followed by hydrolysis to prepare boronic acid. It is also used as a precursor of borate esters, which finds application in the Suzuki coupling reaction. It is an intermediate in the preparation of sodium borohydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only