Titanium(IV) chloride, 99.99% (metals basis)
Titanium(IV) chloride, 99.99% (metals basis)
Titanium(IV) chloride, 99.99% (metals basis)
Titanium(IV) chloride, 99.99% (metals basis)
Thermo Scientific Chemicals

Titanium(IV) chloride, 99.99% (metals basis)

CAS: 7550-45-0 | Cl4Ti | 189.667 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
25 g
100 g
Catalog number 022979.22
also known as 022979-22
Price (USD)
243.00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
243.00
Each
Add to cart
Chemical Identifiers
CAS7550-45-0
IUPAC Nametitanium(4+) tetrachloride
Molecular FormulaCl4Ti
InChI KeyXJDNKRIXUMDJCW-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Titanium(IV) chloride is used to make titanium metal, pigment titanium dioxide and is used in smoke screens. It is involved in the activation of pyrrolidines for better conversion through a modified Bouveault reaction, to the corresponding alpha, alpha -dimethylamines. In organometllic chemistry, it is used to prepare complexes like titanocene dichloride, which is a precursor to Ziegler-Natta catalysts. Tebbe’s reagent is an aluminum containing derivative of titanocene used for the olefination reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only