Cobalt(II) phosphate octahydrate
Cobalt(II) phosphate octahydrate
Cobalt(II) phosphate octahydrate
Thermo Scientific Chemicals

Cobalt(II) phosphate octahydrate

CAS: 10294-50-5 | Co3H16O16P2 | 510.859 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 023111.30
also known as 023111-30
Price (USD)
497.65
Special offer
Online exclusive
Ends: 30-Jun-2026
585.00
Save 87.35 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
497.65
Special offer
Online exclusive
Ends: 30-Jun-2026
585.00
Save 87.35 (15%)
Each
Chemical Identifiers
CAS10294-50-5
IUPAC Nametris(λ²-cobalt(2+)) octahydrate diphosphate
Molecular FormulaCo3H16O16P2
InChI KeyQOQSIXJUYVUEMP-UHFFFAOYSA-H
SMILESO.O.O.O.O.O.O.O.[Co++].[Co++].[Co++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Purple
Assay from Supplier's CofA≥95.0%
It is used as a pigment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only