Cobalt(II) tetrafluoroborate hydrate, 96%
Cobalt(II) tetrafluoroborate hydrate, 96%
Cobalt(II) tetrafluoroborate hydrate, 96%
Thermo Scientific Chemicals

Cobalt(II) tetrafluoroborate hydrate, 96%

CAS: 26490-63-1
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 023142.14
also known as 023142-14
Price (USD)
64.10
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
64.10
Each
Chemical Identifiers
CAS1260029-47-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red-orange to red to brown
FormPowder
Assay (unspecified)≥95.0%
It is a cobalt salt. It is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only