Product Image
Thermo Scientific™

DRI™ Ethyl Alcohol Drugs of Abuse Calibrators

Choose Thermo Scientific™ DRI™ Ethyl Alcohol Drugs of Abuse Calibrators to deliver high analytical accuracy while eliminating time-consuming steps in reagent preparation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
02411 x 5 mL
14061 x 25 mL
Catalog number 0241
Price (USD)
-
Quantity:
1 x 5 mL
Choose Thermo Scientific™ DRI™ Ethyl Alcohol Drugs of Abuse Calibrators to deliver high analytical accuracy while eliminating time-consuming steps in reagent preparation. Ready-to-use in liquid form, these assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration100 mg/dL
DescriptionDRI Alcohol Drugs of Abuse Calibrators
Packaging1 Bottle
Quantity1 x 5 mL
TypeCalibrator
Detectable AnalytesAlcohol
Sample TypePlasma, Serum, Urine
Unit SizeEach
Contents & Storage
2°C to 8°C